3-Amino-4-brom-1H-pyrazol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Bromo-1H-pyrazol-5-amine, 4-Bromo-1H-pyrazol-3-amine, 3-Amino-4-bromopyrazole

AS30564G1 AS30564G5
ACTIAS30564G1EA 0 CHF
ACTIAS30564G1 ACTIAS30564G5
3-Amino-4-brom-1H-pyrazol
3-Amino-4-bromo-1H-pyrazole
Formel: C₃H₄BrN₃
MW: 161,99 g/mol
MDL Number: MFCD00082728
CAS-Nummer: 16461-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR