3,3-Dimethylbutyraldehyd

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3,3-Dimethylbutanal

AS34550G1 AS34550G5
ACTIAS34550G1EA 0 CHF
ACTIAS34550G1 ACTIAS34550G5
3,3-Dimethylbutyraldehyd
3,3-Dimethylbutyraldehyd
Formel: C₆H₁₂O
MW: 100,16 g/mol
MDL Number: MFCD00042807
CAS-Nummer: 2987-16-8
EINECS: 221-054-3
UN: 1989
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR