2-Amino-4-chlorpyrimidin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-chloropyrimidin-2-amine

AS30437G1 AS30437G5
ACTIAS30437G1EA 0 CHF
ACTIAS30437G1 ACTIAS30437G5
2-Amino-4-chlorpyrimidin
2-Amino-4-chloropyrimidine
Formel: C₄H₄ClN₃
MW: 129,55 g/mol
MDL Number: MFCD00038021
CAS-Nummer: 3993-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR