3-Methyl-1H-pyrazol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Methylpyrazole

AS20323G100 AS20323G25
ACTIAS20323G100EA 0 CHF
ACTIAS20323G100 ACTIAS20323G25
3-Methyl-1H-pyrazol
3-Methyl-1H-pyrazole
Formel: C₄H₆N₂
MDL Number: MFCD00005240
CAS-Nummer: 1453-58-3
EINECS: 215-925-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR