2-Chlor-5-fluorpyrimidin
Lieferant: ACTIVATE SCIENTIFIC
AS50273G100
AS50273G5
ACTIAS50273G100EA
0
CHF
ACTIAS50273G100
ACTIAS50273G5
2-Chlor-5-fluorpyrimidin
2-Chloro-5-fluoropyrimidine
Formel:
C₄H₂ClFN₂ MW: 132,52 g/mol |
MDL Number:
MFCD03788197 CAS-Nummer: 62802-42-0 UN: 3267 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...