4,6-Dichlor-5-methylpyrimidin

Lieferant: ACTIVATE SCIENTIFIC

AS23068G1 AS23068G25 AS23068G5
ACTIAS23068G1EA 37 CHF
ACTIAS23068G1 ACTIAS23068G25 ACTIAS23068G5
4,6-Dichlor-5-methylpyrimidin
4,6-Dichloro-5-methylpyrimidine
Formel: C₅H₄Cl₂N₂
MW: 163,01 g/mol
MDL Number: MFCD01646112
CAS-Nummer: 4316-97-6
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR