2-Chlor-5-ethylpyrimidin

Lieferant: ACTIVATE SCIENTIFIC

AS33678G1 AS33678G5
ACTIAS33678G1EA 0 CHF
ACTIAS33678G1 ACTIAS33678G5
2-Chlor-5-ethylpyrimidin
2-Chloro-5-ethylpyrimidine
Formel: C₆H₇ClN₂
MW: 142,59 g/mol
MDL Number: MFCD00799503
CAS-Nummer: 111196-81-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR