2,3-Dimethylhydrochinon
Lieferant: ACTIVATE SCIENTIFIC
AS31379G1
AS31379G5
ACTIAS31379G1EA
0
CHF
ACTIAS31379G1
ACTIAS31379G5
2,3-Dimethylhydrochinon
2,3-Dimethylhydrochinon
Formel:
C₈H₁₀O₂ |
MDL Number:
MFCD00009997 CAS-Nummer: 608-43-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...