5-Brom-2-methylbenzothiazol

Lieferant: ACTIVATE SCIENTIFIC

AS32186G1 AS32186G5
ACTIAS32186G1EA 0 CHF
ACTIAS32186G1 ACTIAS32186G5
5-Brom-2-methylbenzothiazol
5-Bromo-2-methylbenzothiazole
Formel: C₈H₆BrNS
MDL Number: MFCD00603096
CAS-Nummer: 63837-11-6
EINECS: 264-510-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR