2-Adamantanon
Lieferant: ACTIVATE SCIENTIFIC
AS22036G100
AS22036G25
ACTIAS22036G100EA
0
CHF
ACTIAS22036G100
ACTIAS22036G25
2-Adamantanon
2-Adamantanon
Formel:
C₁₀H₁₄O Siedepunkt: 228 °C (1013 hPa) Melting Pt: 43 °C Dichte: 1 g/cm³ (20 °C) Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00074737 CAS-Nummer: 700-58-3 EINECS: 211-847-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...