2,6-Di-tert-butyl-4-methylpyridin

Lieferant: ACTIVATE SCIENTIFIC
Achtung

Synonyms: 2,6-Di-Tert-butyl-4-methylpyridine

AS23347G25 AS23347G5
ACTIAS23347G25EA 0 CHF
ACTIAS23347G25 ACTIAS23347G5
2,6-Di-tert-butyl-4-methylpyridin
2,6-Di-tert-butyl-4-methylpyridin
Formel: C₁₄H₂₃N
MW: 205,34 g/mol
Lager Temperatur: Kühlschrank
MDL Number: MFCD00006305
CAS-Nummer: 38222-83-2
EINECS: 253-834-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR