3-Nitro-2,6-lutidin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2,6-Dimethyl-3-nitropyridine, 3-Nitro-2,6-lutidine

AS32719G1 AS32719G5
ACTIAS32719G1EA 0 CHF
ACTIAS32719G1 ACTIAS32719G5
3-Nitro-2,6-lutidin
3-Nitro-2,6-lutidin
Formel: C₇H₈N₂O₂
MW: 152,15 g/mol
MDL Number: MFCD00023503
CAS-Nummer: 15513-52-7
EINECS: 239-543-5
UN: 1325
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR