2,3-Dichlorpyridin

Lieferant: ACTIVATE SCIENTIFIC
Achtung

AS20093G100 AS20093G25
ACTIAS20093G100EA 0 CHF
ACTIAS20093G100 ACTIAS20093G25
2,3-Dichlorpyridin
2,3-Dichloropyridine
Formel: C₅H₃Cl₂N
Siedepunkt: 202.5…203.5 °C (1013 hPa)
Melting Pt: 70…71 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00006229
CAS-Nummer: 2402-77-9
EINECS: 219-281-8
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR