2,3,5,6-Tetrachlorpyridin
Lieferant: ACTIVATE SCIENTIFIC
|
Gefahr
|
AS24392G100
AS24392G25
ACTIAS24392G100EA
188
CHF
ACTIAS24392G100
ACTIAS24392G25
2,3,5,6-Tetrachlorpyridin
2,3,5,6-Tetrachloropyridine
Formel:
C₅HCl₄N MW: 216,88 g/mol Siedepunkt: 251.6 °C (1013 hPa) Melting Pt: 88…89 °C |
MDL Number:
MFCD00092428 CAS-Nummer: 2402-79-1 EINECS: 219-283-9 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...