2,3-Dibrompyridin

Lieferant: ACTIVATE SCIENTIFIC
Achtung

AS4036G1 AS4036G5
ACTIAS4036G1EA 0 CHF
ACTIAS4036G1 ACTIAS4036G5
2,3-Dibrompyridin
2,3-Dibromopyridine
Formel: C₅H₃Br₂N
MW: 236.89 g/mol
Siedepunkt: 249…249.5 °C (1013 hPa)
Melting Pt: 58…59 °C
MDL Number: MFCD00234014
CAS-Nummer: 13534-89-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR