5-Bromisochinolin
Lieferant: ACTIVATE SCIENTIFIC GMBH
|
Achtung
|
AS7135G100
AS7135G5
ACTIAS7135G100EA
0
CHF
ACTIAS7135G100
ACTIAS7135G5
5-Bromisochinolin
5-Bromoisoquinoline
Formel:
C₉H₆BrN |
MDL Number:
MFCD01646405 CAS-Nummer: 34784-04-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...