6-Nitro-1-indanon

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 6-Nitro-2,3-dihydro-1H-inden-1-one

AS1240G1 AS1240G5
ACTIAS1240G1EA 0 CHF
ACTIAS1240G1 ACTIAS1240G5
6-Nitro-1-indanon
6-Nitro-1-indanon
Formel: C₉H₇NO₃
MW: 177,16 g/mol
MDL Number: MFCD06656903
CAS-Nummer: 24623-24-3
EINECS: 246-366-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR