2,6-Dichlor-4-nitropyridin

Lieferant: ACTIVATE SCIENTIFIC

AS35353G1 AS35353G10 AS35353G5
ACTIAS35353G1EA 0 CHF
ACTIAS35353G1 ACTIAS35353G10 ACTIAS35353G5
2,6-Dichlor-4-nitropyridin
2,6-Dichloro-4-nitropyridine
Formel: C₅H₂Cl₂N₂O₂
MW: 192,99 g/mol
MDL Number: MFCD05670545
CAS-Nummer: 25194-01-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR