Methyl-5-fluornicotinat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 5-Fluoro-nicotinic acid methyl ester, Methyl 5-fluoro-3-pyridinecarboxylate

AS12555G25 AS12555G5
ACTIAS12555G25EA 0 CHF
ACTIAS12555G25 ACTIAS12555G5
Methyl-5-fluornicotinat
Methyl-5-fluornicotinat
Formel: C₇H₆FNO₂
MDL Number: MFCD01692049
CAS-Nummer: 455-70-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR