Methyl-2-fluornicotinat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Methyl 2-fluoronicotinate, 2-Fluoronicotinic acid methyl ester

AS31107G1 AS31107G5
ACTIAS31107G1EA 0 CHF
ACTIAS31107G1 ACTIAS31107G5
Methyl-2-fluornicotinat
Methyl-2-fluornicotinat
Formel: C₇H₆FNO₂
MW: 155,13 g/mol
MDL Number: MFCD03095072
CAS-Nummer: 446-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR