5-Nitropicolinsäure
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
5-Nitro-2-pyridinecarboxylic acid, 5-Nitropyridine-2-carboxylic acid, 5-Nitropicolinic acid
AS1373G1
AS1373G5
ACTIAS1373G1EA
0
CHF
ACTIAS1373G1
ACTIAS1373G5
5-Nitropicolinsäure
5-Nitropicolinsäure
Formel:
C₆H₄N₂O₄ MW: 168,11 g/mol |
MDL Number:
MFCD04114182 CAS-Nummer: 30651-24-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...