Methyl-4-aminonicotinat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Methyl 4-aminonicotinate

AS4031G1 AS4031G5
ACTIAS4031G1EA 0 CHF
ACTIAS4031G1 ACTIAS4031G5
Methyl-4-aminonicotinat
Methyl-4-aminonicotinat
Formel: C₇H₈N₂O₂
MW: 152,15 g/mol
MDL Number: MFCD00955996
CAS-Nummer: 16135-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR