Methyl-6-bromnicotinat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Methyl 6-bromonicotinate

AS33482G1 AS33482G5
ACTIAS33482G1EA 0 CHF
ACTIAS33482G1 ACTIAS33482G5
Methyl-6-bromnicotinat
Methyl-6-bromnicotinat
Formel: C₇H₆BrNO₂
MW: 216.03 g/mol
MDL Number: MFCD04972371
CAS-Nummer: 26218-78-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR