Methyl-5-brom-6-chlornicotinat
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
Methyl 5-bromo-6-chloronicotinate
AS22709G1
AS22709G25
AS22709G5
ACTIAS22709G1EA
15
CHF
ACTIAS22709G1
ACTIAS22709G25
ACTIAS22709G5
Methyl-5-brom-6-chlornicotinat
Methyl-5-brom-6-chlornicotinat
Formel:
C₇H₅BrClNO₂ MW: 250,48 g/mol |
MDL Number:
MFCD03844855 CAS-Nummer: 78686-77-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...