4-Nitroisatosäureanhydrid
Lieferant: ACTIVATE SCIENTIFIC GMBH
ACTIAS36763G1EA
0
CHF
ACTIAS36763G1
ACTIAS36763MG250
4-Nitroisatosäureanhydrid
4-Nitroisatosäureanhydrid
Formel:
C₈H₄N₂O₅ MW: 208,13 g/mol |
MDL Number:
MFCD00798220 CAS-Nummer: 63480-10-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...