Natriummethansulfinat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Methanesulphinic acid, sodium salt

AS20409G100 AS20409G25
ACTIAS20409G100EA 0 CHF
ACTIAS20409G100 ACTIAS20409G25
Natriummethansulfinat
Natriummethansulfinat
Formel: CH₃NaO₂S
MW: 102.09 g/mol
MDL Number: MFCD00040392
CAS-Nummer: 20277-69-4
EINECS: 243-669-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR