Dimethyl-3-oxoglutarat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Dimethyl 1,3-acetonedicarboxylate, Dimethyl acetone-1,3-dicarboxylate

AS34182G10 AS34182G100
ACTIAS34182G10EA 0 CHF
ACTIAS34182G10 ACTIAS34182G100
Dimethyl-3-oxoglutarat
Dimethyl-3-oxoglutarat
Formel: C₇H₁₀O₅
MW: 174,15 g/mol
MDL Number: MFCD00008462
CAS-Nummer: 1830-54-2
EINECS: 217-385-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR