5-Chlor-3-pyridinboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 5-Chloropyridine-3-boronic acid, (5-Chloropyridin-3-yl)boronic acid

AS50718G1 AS50718G5
ACTIAS50718G1EA 0 CHF
ACTIAS50718G1 ACTIAS50718G5
5-Chlor-3-pyridinboronsäure
5-Chloro-3-pyridineboronic acid
Formel: C₅H₅BClNO₂
MW: 157.36 g/mol
MDL Number: MFCD06798243
CAS-Nummer: 872041-85-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR