5-Chlor-2-pyridinboronsäure

Lieferant: ACTIVATE SCIENTIFIC

AS30968G1 AS30968G5
ACTIAS30968G1EA 0 CHF
ACTIAS30968G1 ACTIAS30968G5
5-Chlor-2-pyridinboronsäure
5-Chloro-2-pyridineboronic acid
Formel: C₅H₅BClNO₂
MW: 157.36 g/mol
MDL Number: MFCD07368862
CAS-Nummer: 652148-91-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR