3-Chlor-4-pyridinboronsäure

Lieferant: ACTIVATE SCIENTIFIC

AS51142G1 AS51142G5
ACTIAS51142G1EA 0 CHF
ACTIAS51142G1 ACTIAS51142G5
3-Chlor-4-pyridinboronsäure
3-Chloro-4-pyridineboronic acid
Formel: C₅H₅BClNO₂
MW: 157.36 g/mol
MDL Number: MFCD03425942
CAS-Nummer: 458532-98-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR