2,6-Dichlor-3-pyridinboronsäure
Lieferant: ACTIVATE SCIENTIFIC
AS50920G1
AS50920G5
ACTIAS50920G1EA
0
CHF
ACTIAS50920G1
ACTIAS50920G5
2,6-Dichlor-3-pyridinboronsäure
2,6-Dichloro-3-pyridineboronic acid
Formel:
C₅H₄BCl₂NO₂ |
MDL Number:
MFCD03094989 CAS-Nummer: 148493-34-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...