2,5-Dichlor-4-pyridinboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2,5-Dichloropyridine-4-boronic acid, 2,5-Dichloro-4-pyridylboronic acid

AS51551G1 AS51551G5
ACTIAS51551G1EA 0 CHF
ACTIAS51551G1 ACTIAS51551G5
2,5-Dichlor-4-pyridinboronsäure
2,5-Dichloro-4-pyridineboronic acid
Formel: C₅H₄BCl₂NO₂
MDL Number: MFCD06798254
CAS-Nummer: 847664-64-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR