4-Methylpicolinsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Methylpicolinic acid, 4-Methylpyridine-2-carboxylic acid

AS31952G1 AS31952G5
ACTIAS31952G1EA 0 CHF
ACTIAS31952G1 ACTIAS31952G5
4-Methylpicolinsäure
4-Methylpicolinsäure
Formel: C₇H₇NO₂
MW: 137.14 g/mol
MDL Number: MFCD03731175
CAS-Nummer: 4021-08-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR