3-Methylpicolinsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Methylpyridine-2-carboxylic acid

AS1580G100 AS1580G5
ACTIAS1580G100EA 0 CHF
ACTIAS1580G100 ACTIAS1580G5
3-Methylpicolinsäure
3-Methylpicolinsäure
Formel: C₇H₇NO₂
MW: 137.14 g/mol
MDL Number: MFCD00191553
CAS-Nummer: 4021-07-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR