4-Bromnicotinonitril

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Bromo-3-cyanopyridine

AS30595G1 AS30595G5
ACTIAS30595G1EA 0 CHF
ACTIAS30595G1 ACTIAS30595G5
4-Bromnicotinonitril
4-Bromnicotinonitril
Formel: C₆H₃BrN₂
MW: 183,01 g/mol
MDL Number: MFCD08686971
CAS-Nummer: 154237-70-4
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR