4-Brom-2-pyridincarbonitril

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Bromo-2-cyanopyridine

AS31937G1 AS31937G5
ACTIAS31937G1EA 0 CHF
ACTIAS31937G1 ACTIAS31937G5
4-Brom-2-pyridincarbonitril
4-Brom-2-pyridincarbonitril
Formel: C₆H₃BrN₂
MDL Number: MFCD04065805
CAS-Nummer: 62150-45-2
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR