3-Fluorisonicotinsäure
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
3-Fluoropyridine-4-carboxylic acid
AS31274G1
AS31274G5
ACTIAS31274G1EA
0
CHF
ACTIAS31274G1
ACTIAS31274G5
3-Fluorisonicotinsäure
3-Fluorisonicotinsäure
Formel:
C₆H₄FNO₂ MW: 141,1 g/mol |
MDL Number:
MFCD03789251 CAS-Nummer: 393-53-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...