3-Fluorpicolinsäure
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
3-fluoropyridine-2-carboxylic acid, 3-Fluoro-2-pyridinecarboxylic acid
AS31774G1
AS31774G5
ACTIAS31774G1EA
0
CHF
ACTIAS31774G1
ACTIAS31774G5
3-Fluorpicolinsäure
3-Fluorpicolinsäure
Formel:
C₆H₄FNO₂ MW: 141,1 g/mol |
MDL Number:
MFCD04114108 CAS-Nummer: 152126-31-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...