N-Methylphenethylamin

Lieferant: ACTIVATE SCIENTIFIC

AS20375G25 AS20375G5
ACTIAS20375G25EA 158 CHF
ACTIAS20375G25 ACTIAS20375G5
N-Methylphenethylamin
N-Methylphenethylamin
Formel: C₉H₁₃N
MW: 135,21 g/mol
MDL Number: MFCD00008291
CAS-Nummer: 589-08-2
EINECS: 209-632-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR