5-Chlorpicolinsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 5-Chloro-2-pyridinecarboxylic acid, 5-Chloropyridine-2-carboxylic acid, 5-Chloro-2-picolinic acid

AS22679G100 AS22679G25 AS22679G5
ACTIAS22679G100EA 290 CHF
ACTIAS22679G100 ACTIAS22679G25 ACTIAS22679G5
5-Chlorpicolinsäure
5-Chlorpicolinsäure
Formel: C₆H₄ClNO₂
MW: 157.56 g/mol
MDL Number: MFCD04114192
CAS-Nummer: 86873-60-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR