4-Chlorisatin
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
4-Chloroindoline-2,3-dione
AS21783G100
AS21783G25
AS21783G5
ACTIAS21783G100EA
251
CHF
ACTIAS21783G100
ACTIAS21783G25
ACTIAS21783G5
4-Chlorisatin
4-Chlorisatin
Formel:
C₈H₄ClNO₂ MW: 181,58 g/mol |
MDL Number:
MFCD00129178 CAS-Nummer: 6344-05-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...