3-Fluor-5-formylpyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 5-Fluoronicotinaldehyde

AS31948G1 AS31948G5
ACTIAS31948G1EA 0 CHF
ACTIAS31948G1 ACTIAS31948G5
3-Fluor-5-formylpyridin
3-Fluor-5-formylpyridin
Formel: C₆H₄FNO
MW: 125,1 g/mol
MDL Number: MFCD07778429
CAS-Nummer: 39891-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR