3-Fluor-2-formylpyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Fluoropyridine-2-carboxaldehyde, 3-Fluoropicolinaldehyde, 3-Fluoropyridine-2-carbaldehyde, 3-Fluoro-2-pyridinecarboxaldehyde

AS31240G1 AS31240G5
ACTIAS31240G1EA 0 CHF
ACTIAS31240G1 ACTIAS31240G5
3-Fluor-2-formylpyridin
3-Fluor-2-formylpyridin
Formel: C₆H₄FNO
MDL Number: MFCD07781234
CAS-Nummer: 31224-43-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR