4-Formyl-2-methoxypyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Methoxypyridine-4-carbaldehyde, 2-Methoxyisonicotinaldehyde

AS1058G1 AS1058G5
ACTIAS1058G1EA 0 CHF
ACTIAS1058G1 ACTIAS1058G5
4-Formyl-2-methoxypyridin
4-Formyl-2-methoxypyridin
Formel: C₇H₇NO₂
MDL Number: MFCD06410680
CAS-Nummer: 72716-87-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR