3-Formyl-2-methoxypyridin
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
2-Methoxynicotinaldehyde, 2-Methoxy-3-pyridinecarboxaldehyde, 2-Methoxy-pyridine-3-carbaldehyde, 2-Methoxypyridine-3-carboxaldehyde
AS50971G1
AS50971G5
ACTIAS50971G1EA
0
CHF
ACTIAS50971G1
ACTIAS50971G5
3-Formyl-2-methoxypyridin
3-Formyl-2-methoxypyridin
Formel:
C₇H₇NO₂ |
MDL Number:
MFCD04115112 CAS-Nummer: 71255-09-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...