3-Chlor-2-formylpyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Chloropyridine-2-carboxaldehyde

AS35640G1 AS35640G5
ACTIAS35640G1EA 76 CHF
ACTIAS35640G1 ACTIAS35640G5
3-Chlor-2-formylpyridin
3-Chlor-2-formylpyridin
Formel: C₆H₄ClNO
MW: 141,56 g/mol
MDL Number: MFCD09991855
CAS-Nummer: 206181-90-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR