2-Chlor-3-formylpyridin
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
2-Chloropyridine-3-carboxaldehyde, 2-Chloronicotinaldehyde, 2-Chloro-3-pyridinecarboxaldehyde
AS50672G100
AS50672G5
ACTIAS50672G100EA
0
CHF
ACTIAS50672G100
ACTIAS50672G5
2-Chlor-3-formylpyridin
2-Chlor-3-formylpyridin
Formel:
C₆H₄ClNO MW: 141,56 g/mol |
MDL Number:
MFCD01315308 CAS-Nummer: 36404-88-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...