5-Brom-2-formylpyridin
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
5-Bromopyridine-2-carbaldehyde, 5-Bromopicolinaldehyde, 5-Bromopyridine-2-carboxaldehyde
AS20598G1
AS20598G25
AS20598G5
ACTIAS20598G1EA
21
CHF
ACTIAS20598G1
ACTIAS20598G25
ACTIAS20598G5
5-Brom-2-formylpyridin
5-Brom-2-formylpyridin
Formel:
C₆H₄BrNO MW: 186,01 g/mol |
MDL Number:
MFCD04112535 CAS-Nummer: 31181-90-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...