3-Brom-4-formylpyridin
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
3-Bromo-4-pyridinecarboxaldehyde, 3-Bromoisonicotinaldehyde, 3-Bromopyridine-4-carboxaldehyde
AS51043G1
AS51043G5
ACTIAS51043G1EA
37
CHF
ACTIAS51043G1
ACTIAS51043G5
3-Brom-4-formylpyridin
3-Brom-4-formylpyridin
Formel:
C₆H₄BrNO MW: 186,01 g/mol |
MDL Number:
MFCD05864506 CAS-Nummer: 70201-43-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...