2-Brom-3-formylpyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Bromopyridine-3-carboxaldehyde, 2-Bromonicotinaldehyde, 2-Bromo-3-pyridinecarboxaldehyde

AS40197G10 AS40197G5
ACTIAS40197G10EA 0 CHF
ACTIAS40197G10 ACTIAS40197G5
2-Brom-3-formylpyridin
2-Brom-3-formylpyridin
Formel: C₆H₄BrNO
MW: 186,01 g/mol
MDL Number: MFCD04966945
CAS-Nummer: 128071-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR